siRNA Details
| Gene Symbol: F2rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053897.2 | NP_446349.1 | 2 | 916 | ||
| GenBank mRNA | BC099765.1 | - | 2 | 916 | 7930573 | |
| U61373.1 | - | 2 | 829 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F2rl1| Gene Symbol: F2rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053897.2 | NP_446349.1 | 2 | 916 | ||
| GenBank mRNA | BC099765.1 | - | 2 | 916 | 7930573 | |
| U61373.1 | - | 2 | 829 | |||