siRNA Details
Gene Symbol: Oplah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053904.1 | NP_446356.1 | 16 | 2231 | ||
XM_006241771.2 | XP_006241833.1 | 16 | 3891 | |||
XM_006241772.2 | XP_006241834.1 | 16 | 3891 | |||
XM_006241773.2 | XP_006241835.1 | 16 | 3774 | |||
XM_006241774.2 | XP_006241836.1 | 16 | 2569 | |||
XM_006241775.2 | XP_006241837.1 | 17 | 2428 | |||
GenBank mRNA | BC085330.1 | - | 16 | 2296 | 7096390 | |
U70825.1 | - | 16 | 2231 |
