siRNA Details
| Gene Symbol: Oplah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053904.1 | NP_446356.1 | 8 | 1105 | ||
| XM_006241771.2 | XP_006241833.1 | 8 | 2765 | |||
| XM_006241772.2 | XP_006241834.1 | 8 | 2765 | |||
| XM_006241773.2 | XP_006241835.1 | 8 | 2648 | |||
| XM_006241774.2 | XP_006241836.1 | 8 | 1443 | |||
| XM_006241775.2 | XP_006241837.1 | 9 | 1302 | |||
| GenBank mRNA | BC085330.1 | - | 8 | 1170 | 7096390 | |
| U70825.1 | - | 8 | 1105 | |||

Oplah
Genomic Map