siRNA Details
| Gene Symbol: Gsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053906.2 | NP_446358.2 | 11 | 955 | ||
| GenBank mRNA | CK357934.1 | - | 4 | 418 | ||
| U73174.1 | - | 1094 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gsr| Gene Symbol: Gsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053906.2 | NP_446358.2 | 11 | 955 | ||
| GenBank mRNA | CK357934.1 | - | 4 | 418 | ||
| U73174.1 | - | 1094 | ||||