siRNA Details
| Gene Symbol: Capn9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271140.1 | NP_001258069.1 | 10 | 1159 | ||
| GenBank mRNA | U89514.1 | - | 10 | 1074 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Capn9| Gene Symbol: Capn9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271140.1 | NP_001258069.1 | 10 | 1159 | ||
| GenBank mRNA | U89514.1 | - | 10 | 1074 | ||