siRNA Details
Gene Symbol: Capn9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271140.1 | NP_001258069.1 | 16, 17 | 1817 | ||
GenBank mRNA | U89514.1 | - | 16, 17 | 1738 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Capn9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271140.1 | NP_001258069.1 | 16, 17 | 1817 | ||
GenBank mRNA | U89514.1 | - | 16, 17 | 1738 |