siRNA Details
| Gene Symbol: Pabpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135008.1 | NP_001128480.1 | 2 | 433 | ||
| XM_008770659.1 | XP_008768881.1 | 3 | 2088 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pabpn1| Gene Symbol: Pabpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135008.1 | NP_001128480.1 | 2 | 433 | ||
| XM_008770659.1 | XP_008768881.1 | 3 | 2088 | |||