siRNA Details
| Gene Symbol: Inpp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053917.1 | NP_446369.1 | 6 | 691 | ||
| XM_008772462.1 | XP_008770684.1 | 6 | 534 | |||
| XM_008772463.1 | XP_008770685.1 | 6 | 534 | |||
| XM_008772464.1 | XP_008770686.1 | 6 | 691 | |||
| XM_008772465.1 | XP_008770687.1 | 6 | 534 | |||
| GenBank mRNA | U96920.1 | - | 6 | 691 | ||
| U96921.1 | - | 6 | 691 | |||

Inpp4b
Genomic Map