siRNA Details
| Gene Symbol: Abcc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053924.1 | NP_446376.1 | 22 | 3302 | ||
| XM_006248593.2 | XP_006248655.1 | 23 | 3356 | |||
| GenBank mRNA | AB020209.1 | - | 22 | 3302 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcc5| Gene Symbol: Abcc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053924.1 | NP_446376.1 | 22 | 3302 | ||
| XM_006248593.2 | XP_006248655.1 | 23 | 3356 | |||
| GenBank mRNA | AB020209.1 | - | 22 | 3302 | ||