siRNA Details
| Gene Symbol: Pip4k2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053926.2 | NP_446378.1 | 7 | 1097 | ||
| GenBank mRNA | AB032899.1 | - | 7 | 1097 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pip4k2a| Gene Symbol: Pip4k2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053926.2 | NP_446378.1 | 7 | 1097 | ||
| GenBank mRNA | AB032899.1 | - | 7 | 1097 | ||