siRNA Details
| Gene Symbol: Slc7a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053929.1 | NP_446381.1 | 11 | 1183 | ||
| XM_006228875.2 | XP_006228937.1 | 12 | 1285 | |||
| XM_006228876.2 | XP_006228938.1 | 12 | 1665 | |||
| XM_006228877.2 | XP_006228939.1 | 11 | 1544 | |||
| XM_006228878.2 | XP_006228940.1 | 8 | 850 | |||
| XM_008759103.1 | XP_008757325.1 | 11 | 1569 | |||
| XM_008759104.1 | XP_008757326.1 | 11 | 1224 | |||
| GenBank mRNA | AB029559.1 | - | 11 | 1183 | ||
| BC081750.1 | - | 10 | 1599 | |||
| BC098909.1 | - | 11 | 1173 | 7109606 | ||

Slc7a9
Genomic Map