siRNA Details
| Gene Symbol: B3gnt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053932.1 | NP_446384.1 | 4 | 929 | ||
| XM_008768849.1 | XP_008767071.1 | 2 | 666 | |||
| GenBank mRNA | FQ225758.1 | - | 1 | 404 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B3gnt5| Gene Symbol: B3gnt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053932.1 | NP_446384.1 | 4 | 929 | ||
| XM_008768849.1 | XP_008767071.1 | 2 | 666 | |||
| GenBank mRNA | FQ225758.1 | - | 1 | 404 | ||