siRNA Details
| Gene Symbol: Pcdha13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053934.2 | NP_446386.2 | 1 | 2294 | ||
| GenBank mRNA | AB045586.1 | - | 1 | 1572 | ||
| AB113396.1 | - | 1 | 2294 | |||
| AY573975.1 | - | 1 | 2294 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdha13| Gene Symbol: Pcdha13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053934.2 | NP_446386.2 | 1 | 2294 | ||
| GenBank mRNA | AB045586.1 | - | 1 | 1572 | ||
| AB113396.1 | - | 1 | 2294 | |||
| AY573975.1 | - | 1 | 2294 | |||