siRNA Details
| Gene Symbol: Cdh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255559.2 | XP_006255621.1 | 4 | 840 | ||
| GenBank mRNA | FQ220836.1 | - | 4 | 350 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh3| Gene Symbol: Cdh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255559.2 | XP_006255621.1 | 4 | 840 | ||
| GenBank mRNA | FQ220836.1 | - | 4 | 350 | ||