siRNA Details
| Gene Symbol: Kcnh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053949.1 | NP_446401.1 | 6 | 1303 | ||
| XM_006235867.2 | XP_006235929.1 | 6 | 1436 | |||
| XM_008762630.1 | XP_008760852.1 | 2 | 624 | |||
| GenBank mRNA | AY669863.1 | - | 2 | 277 | ||
| U75210.1 | - | 1 | 79 | |||
| Z96106.1 | - | 6 | 1303 | |||

Kcnh2
Genomic Map