siRNA Details
Gene Symbol: Crym | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053955.1 | NP_446407.1 | 6 | 799 | ||
GenBank mRNA | BC088121.1 | - | 6 | 830 | 7323247 | |
Y17328.1 | - | 6 | 799 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Crym | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053955.1 | NP_446407.1 | 6 | 799 | ||
GenBank mRNA | BC088121.1 | - | 6 | 830 | 7323247 | |
Y17328.1 | - | 6 | 799 |