siRNA Details
| Gene Symbol: Crym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053955.1 | NP_446407.1 | 4 | 477 | ||
| GenBank mRNA | BC088121.1 | - | 4 | 508 | 7323247 | |
| Y17328.1 | - | 4 | 477 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crym| Gene Symbol: Crym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053955.1 | NP_446407.1 | 4 | 477 | ||
| GenBank mRNA | BC088121.1 | - | 4 | 508 | 7323247 | |
| Y17328.1 | - | 4 | 477 | |||