siRNA Details
| Gene Symbol: Ccr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053958.1 | NP_446410.1 | 3 | 772 | ||
| XM_006244178.2 | XP_006244240.1 | 3 | 1550 | |||
| GenBank mRNA | Y13400.1 | - | 3 | 772 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccr3| Gene Symbol: Ccr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053958.1 | NP_446410.1 | 3 | 772 | ||
| XM_006244178.2 | XP_006244240.1 | 3 | 1550 | |||
| GenBank mRNA | Y13400.1 | - | 3 | 772 | ||