siRNA Details
| Gene Symbol: Ccr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053960.3 | NP_446412.2 | 2 | 779 | ||
| GenBank mRNA | BC078756.1 | - | 1 | 714 | 7109679 | |
| Y12009.1 | - | 2 | 789 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccr5| Gene Symbol: Ccr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053960.3 | NP_446412.2 | 2 | 779 | ||
| GenBank mRNA | BC078756.1 | - | 1 | 714 | 7109679 | |
| Y12009.1 | - | 2 | 789 | |||