siRNA Details
| Gene Symbol: Slc25a20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053965.2 | NP_446417.2 | 6 | 654 | ||
| GenBank mRNA | BC081749.1 | - | 6 | 654 | 7134670 | |
| FQ209681.1 | - | 6 | 610 | |||
| X97831.1 | - | 6 | 611 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a20| Gene Symbol: Slc25a20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053965.2 | NP_446417.2 | 6 | 654 | ||
| GenBank mRNA | BC081749.1 | - | 6 | 654 | 7134670 | |
| FQ209681.1 | - | 6 | 610 | |||
| X97831.1 | - | 6 | 611 | |||