siRNA Details
| Gene Symbol: Nln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053970.2 | NP_446422.2 | 13 | 2136 | ||
| XM_006231881.2 | XP_006231943.1 | 14 | 2368 | |||
| GenBank mRNA | BC072687.1 | - | 13 | 2136 | 7099208 | |
| FQ228111.1 | - | 6 | 843 | |||
| FQ230641.1 | - | 6 | 843 | |||
| X87157.1 | - | 13 | 2244 | |||

Nln
Genomic Map