siRNA Details
| Gene Symbol: Eif4e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053974.2 | NP_446426.1 | 5 | 386 | ||
| XM_008761488.1 | XP_008759710.1 | 6 | 499 | |||
| GenBank mRNA | BC087001.1 | - | 5 | 386 | 7122606 | |
| FQ218941.1 | - | 5 | 378 | |||
| X83399.1 | - | 5 | 421 | |||

Eif4e
Genomic Map