siRNA Details
| Gene Symbol: Rab28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053978.1 | NP_446430.1 | 3, 4 | 281 | ||
| GenBank mRNA | X78606.1 | - | 3, 4 | 281 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab28| Gene Symbol: Rab28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053978.1 | NP_446430.1 | 3, 4 | 281 | ||
| GenBank mRNA | X78606.1 | - | 3, 4 | 281 | ||