siRNA Details
Gene Symbol: Arl5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053979.2 | NP_446431.1 | 1, 2 | 236 | ||
GenBank mRNA | BC097294.1 | - | 1, 2 | 236 | 7458249 | |
X78604.1 | - | 1, 2 | 85 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Arl5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053979.2 | NP_446431.1 | 1, 2 | 236 | ||
GenBank mRNA | BC097294.1 | - | 1, 2 | 236 | 7458249 | |
X78604.1 | - | 1, 2 | 85 |