siRNA Details
Gene Symbol: Arl5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053979.2 | NP_446431.1 | 6 | 717 | ||
XM_008761696.1 | XP_008759918.1 | 6 | 789 | |||
GenBank mRNA | BC097294.1 | - | 6 | 717 | 7458249 | |
X78604.1 | - | 6 | 566 |
