siRNA Details
| Gene Symbol: Kcnj12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053981.2 | NP_446433.2 | 2 | 1442 | ||
| XM_008767763.1 | XP_008765985.1 | 2 | 8732 | |||
| GenBank mRNA | AY376691.1 | - | 1 | 1110 | ||
| BC127487.1 | - | 2 | 1442 | 8364361 | ||
| X78461.1 | - | 2 | 1173 | |||

Kcnj12
Genomic Map