siRNA Details
| Gene Symbol: Calb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053988.1 | NP_446440.1 | 6 | 517 | ||
| XM_008772466.1 | XP_008770688.1 | 7 | 696 | |||
| GenBank mRNA | BC087603.1 | - | 6 | 487 | 7312595 | |
| X66974.1 | - | 6 | 517 | |||

Calb2
Genomic Map