siRNA Details
Gene Symbol: Calb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053988.1 | NP_446440.1 | 8, 9 | 618 | ||
XM_008772466.1 | XP_008770688.1 | 9, 10 | 797 | |||
GenBank mRNA | BC087603.1 | - | 8, 9 | 588 | 7312595 | |
X66974.1 | - | 8, 9 | 618 |
