siRNA Details
| Gene Symbol: Ptpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053990.1 | NP_446442.1 | 6 | 590 | ||
| GenBank mRNA | X58828.1 | - | 6 | 590 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptpn2| Gene Symbol: Ptpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053990.1 | NP_446442.1 | 6 | 590 | ||
| GenBank mRNA | X58828.1 | - | 6 | 590 | ||