siRNA Details
| Gene Symbol: Nlgn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053992.1 | NP_446444.1 | 8 | 2244 | ||
| GenBank mRNA | U41662.1 | - | 8 | 2244 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nlgn2| Gene Symbol: Nlgn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053992.1 | NP_446444.1 | 8 | 2244 | ||
| GenBank mRNA | U41662.1 | - | 8 | 2244 | ||