siRNA Details
Gene Symbol: Kcnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054000.2 | NP_446452.2 | 3 | 1943 | ||
XM_008763474.1 | XP_008761696.1 | 2 | 1101 | |||
GenBank mRNA | M77482.1 | - | 3 | 1946 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kcnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054000.2 | NP_446452.2 | 3 | 1943 | ||
XM_008763474.1 | XP_008761696.1 | 2 | 1101 | |||
GenBank mRNA | M77482.1 | - | 3 | 1946 |