siRNA Details
| Gene Symbol: Scarb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054001.2 | NP_446453.1 | 4 | 839 | ||
| GenBank mRNA | BC061853.1 | - | 4 | 839 | 5621697 | |
| D10587.1 | - | 4 | 778 | |||
| M68965.1 | - | 4 | 787 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scarb2| Gene Symbol: Scarb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054001.2 | NP_446453.1 | 4 | 839 | ||
| GenBank mRNA | BC061853.1 | - | 4 | 839 | 5621697 | |
| D10587.1 | - | 4 | 778 | |||
| M68965.1 | - | 4 | 787 | |||