siRNA Details
| Gene Symbol: Cand1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054004.2 | NP_446456.1 | 13 | 3642 | ||
| GenBank mRNA | D87671.1 | - | 13 | 3644 | ||
| FQ225295.1 | - | 2 | 129 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cand1| Gene Symbol: Cand1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054004.2 | NP_446456.1 | 13 | 3642 | ||
| GenBank mRNA | D87671.1 | - | 13 | 3644 | ||
| FQ225295.1 | - | 2 | 129 | |||