siRNA Details
Gene Symbol: Cand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054004.2 | NP_446456.1 | 10 | 3264 | ||
GenBank mRNA | D87671.1 | - | 10 | 3266 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054004.2 | NP_446456.1 | 10 | 3264 | ||
GenBank mRNA | D87671.1 | - | 10 | 3266 |