siRNA Details
| Gene Symbol: Neu3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054010.1 | NP_446462.1 | 3 | 428 | ||
| XM_008759664.1 | XP_008757886.1 | 2 | 3851 | |||
| XM_008759665.1 | XP_008757887.1 | 3 | 549 | |||
| XM_008759666.1 | XP_008757888.1 | 2 | 347 | |||
| GenBank mRNA | AB026841.1 | - | 3 | 428 | ||
| BC100625.1 | - | 3 | 376 | 7935525 | ||

Neu3
Genomic Map