siRNA Details
| Gene Symbol: Ppp2r2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057116.1 | NP_476457.1 | 83 | |||
| GenBank mRNA | D38261.1 | - | 83 | |||
| Gene Symbol: LOC103693768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770232.1 | XP_008768454.1 | 2 | 443 | ||

Ppp2r2c
Genomic Map