siRNA Details
| Gene Symbol: Gpr176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270986.1 | NP_001257915.1 | 3 | 794 | ||
| GenBank mRNA | D38450.1 | - | 2 | 411 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr176| Gene Symbol: Gpr176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270986.1 | NP_001257915.1 | 3 | 794 | ||
| GenBank mRNA | D38450.1 | - | 2 | 411 | ||
Set Membership: |
|