siRNA Details
| Gene Symbol: Psmc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057122.1 | NP_476463.1 | 6, 7 | 679 | ||
| GenBank mRNA | BC063145.1 | - | 6, 7 | 689 | 6889167 | |
| D50695.1 | - | 6, 7 | 679 | |||
| FQ225557.1 | - | 6, 7 | 704 | |||
| FQ226113.1 | - | 6, 7 | 688 | |||

Psmc4
Genomic Map