siRNA Details
| Gene Symbol: Psmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057123.1 | NP_476464.1 | 7 | 731 | ||
| GenBank mRNA | BC063157.1 | - | 8 | 743 | 6919865 | |
| D50696.1 | - | 7 | 731 | |||
| FQ209916.1 | - | 8 | 736 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmc1| Gene Symbol: Psmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057123.1 | NP_476464.1 | 7 | 731 | ||
| GenBank mRNA | BC063157.1 | - | 8 | 743 | 6919865 | |
| D50696.1 | - | 7 | 731 | |||
| FQ209916.1 | - | 8 | 736 | |||