siRNA Details
| Gene Symbol: Slc26a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057127.1 | NP_476468.1 | 5 | 1799 | ||
| XM_006254784.2 | XP_006254846.1 | 3 | 3775 | |||
| XM_006254785.2 | XP_006254847.1 | 4 | 2445 | |||
| XM_006254786.2 | XP_006254848.1 | 4 | 3253 | |||
| XM_006254787.2 | XP_006254849.1 | 4 | 1793 | |||
| XM_006254788.2 | XP_006254850.1 | 3 | 1658 | |||
| GenBank mRNA | D82883.1 | - | 5 | 1799 | ||

Slc26a2
Genomic Map