siRNA Details
Gene Symbol: Chrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057134.1 | NP_476475.1 | 3 | 315 | ||
XM_006248556.2 | XP_006248618.1 | 3 | 697 | |||
XM_006248557.2 | XP_006248619.1 | 3 | 721 | |||
GenBank mRNA | FQ222446.1 | - | 3 | 354 | ||
FQ223218.1 | - | 3 | 354 |
