siRNA Details
| Gene Symbol: Hnrnpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057141.1 | NP_476482.1 | 1 | 67 | ||
| XM_006253552.2 | XP_006253614.1 | 3 | 474 | |||
| XM_006253553.2 | XP_006253615.1 | 2 | 170 | |||
| XM_006253554.2 | XP_006253616.1 | 3 | 474 | |||
| XM_006253555.2 | XP_006253617.1 | 3 | 474 | |||
| GenBank mRNA | BC061867.1 | - | 2 | 154 | 5598872 | |
| D17711.1 | - | 1 | 67 | |||

Hnrnpk
Genomic Map