siRNA Details
| Gene Symbol: Sec22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057147.2 | NP_476488.2 | 2 | 162 | ||
| XM_006248419.2 | XP_006248481.1 | 3 | 364 | |||
| XM_006248420.2 | XP_006248482.1 | 2 | 399 | |||
| XM_008768692.1 | XP_008766914.1 | 2 | 611 | |||
| XM_008768693.1 | XP_008766915.1 | 2 | 613 | |||
| GenBank mRNA | BC081746.1 | - | 2 | 162 | 7135589 | |

Sec22a
Genomic Map