siRNA Details
| Gene Symbol: Scaper | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766363.1 | XP_008764585.1 | 5 | 403 | ||
| XM_008766364.1 | XP_008764586.1 | 5 | 508 | |||
| XM_008766365.1 | XP_008764587.1 | 5 | 403 | |||
| GenBank mRNA | AF347028.1 | - | 5 | 323 | ||

Scaper
Genomic Map