siRNA Details
| Gene Symbol: Nsmf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057190.2 | NP_476538.2 | 6 | 952 | ||
| XM_006233554.2 | XP_006233616.1 | 6 | 1255 | |||
| XM_008761575.1 | XP_008759797.1 | 5 | 1249 | |||
| XM_008761576.1 | XP_008759798.1 | 5 | 1249 | |||
| GenBank mRNA | AJ293697.1 | - | 6 | 885 | ||
| AJ534641.1 | - | 1 | 37 | |||
| AJ534642.1 | - | 6 | 884 | |||
| BC087719.1 | - | 6 | 913 | 7318793 | ||

Nsmf
Genomic Map