siRNA Details
Gene Symbol: Wipf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057192.2 | NP_476540.2 | 2 | 290 | ||
GenBank mRNA | AJ303456.1 | - | 1 | 123 | ||
BC072475.1 | - | 2 | 290 | 7100751 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wipf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057192.2 | NP_476540.2 | 2 | 290 | ||
GenBank mRNA | AJ303456.1 | - | 1 | 123 | ||
BC072475.1 | - | 2 | 290 | 7100751 |