siRNA Details
| Gene Symbol: Plscr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057194.1 | NP_476542.1 | 3 | 507 | ||
| XM_006243536.1 | XP_006243598.1 | 4 | 582 | |||
| XM_006243537.1 | XP_006243599.1 | 4 | 577 | |||
| XM_006243538.2 | XP_006243600.1 | 3 | 529 | |||
| GenBank mRNA | AY024347.1 | - | 3 | 507 | ||
| BC085715.1 | - | 4 | 506 | 7109899 | ||

Plscr1
Genomic Map