siRNA Details
| Gene Symbol: Plscr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057194.1 | NP_476542.1 | 2, 3 | 178 | ||
| XM_006243538.2 | XP_006243600.1 | 2, 3 | 200 | |||
| GenBank mRNA | AY024347.1 | - | 2, 3 | 178 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plscr1| Gene Symbol: Plscr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057194.1 | NP_476542.1 | 2, 3 | 178 | ||
| XM_006243538.2 | XP_006243600.1 | 2, 3 | 200 | |||
| GenBank mRNA | AY024347.1 | - | 2, 3 | 178 | ||