siRNA Details
| Gene Symbol: Ppat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057198.2 | NP_476546.1 | 7 | 1349 | ||
| GenBank mRNA | BC086999.1 | - | 7 | 1349 | 7189259 | |
| D10853.1 | - | 7 | 857 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppat| Gene Symbol: Ppat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057198.2 | NP_476546.1 | 7 | 1349 | ||
| GenBank mRNA | BC086999.1 | - | 7 | 1349 | 7189259 | |
| D10853.1 | - | 7 | 857 | |||