siRNA Details
Gene Symbol: Ncr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057199.1 | NP_476547.1 | 6 | 713 | ||
GenBank mRNA | AF082533.1 | - | 6 | 713 | ||
AJ012741.1 | - | 6 | 703 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ncr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057199.1 | NP_476547.1 | 6 | 713 | ||
GenBank mRNA | AF082533.1 | - | 6 | 713 | ||
AJ012741.1 | - | 6 | 703 |